missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for SUB-GEL Mini Horizontal Gel Electrophoresis Units, 1.5mm Thick

Product Code. 11813493
Change view
Click to view available options
No. of Wells:
1 Prep, 1 Marker
10
12
16
2 Prep, 2 Marker
4 Prep, 2 Marker
8
Volume (Metric) Sample:
15 μL
17 μL
20 μL
27 μL
37 μL
72 μL
135 μL
304 μL
missing translation for 'unitSize':
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Wells Volume (Metric) Sample missing translation for 'unitSize'
11813493 16 15 μL Each
11803483 10 27 μL Each
11833483 1 Prep, 1 Marker 304 μL Each
11873483 12 20 μL Each
11857553 2 Prep, 2 Marker 135 μL Each
11897553 4 Prep, 2 Marker 72 μL Each
11837563 8 37 μL Each
11877563 8 17 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 missing translation for 'options'
Product Code. 11813493
  • ONLINE EXCLUSIVE PRICE
    £34.15
    £18.19 / Pack of 1
    Save £15.96
    46% Off
Estimated Shipment: 15-09-2026
Add to basket
Add to basket

For use with SUB-GEL Mini Horizontal Gel Electrophoresis Units.

  • Recommended to scale-up nucleic acid volumes for preparatory techniques
  • Maximises sample volume
  • Height-adjustable without any requirement for specialist tools or comb holders, to give user full control over well depth and sample-loading volume
  • Rigid comb back prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single- and multi-channel pipets
  • 8-sample and 12-sample combs are Multichannel pipettor compatible

Specifications

Color Red
For Use With (Equipment) Sub-Gel Mini Horizontal Gel Electrophoresis Unit
Type Comb
No. of Wells 16
Quantity 1 Each
Thickness (Metric) 1.5 mm
Volume (Metric) Sample 15 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.