missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Color-Coded Autoclavable Identification Tape

Instrument marking tape for easy identification and color coding items. Even sticks to PTFE.

Brand:  Fisherbrand™

Additional Details : Weight : 0.18140kg

 View more versions of this product

Product Code. 17867261

  • £5.45 / Each
Estimated Shipment: 31-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Instrument marking tape for easy identification and color coding items. Even sticks to PTFE.

  • Vinyl self-stick tapes color-code autoclave items such as surgical instruments for easy identification
  • Attach securely and safely to any clean surface
  • Resist water, oil, and acids
  • Nontoxic

Order Info

In 16 ft.L x 0.37 in. W (5m x 9.6mm) rolls.

Specifications

Specifications

Autoclavable
Orange
Vinyl
Resist Water, Oil and Acids
16 ft. x 0.37 in.
4.8 m
Self-adhesive
Tape
Identification Tape
1 Ea.
16 ft.
0.37 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers