missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 384-Well Polystyrene Plates

Product Code. 13555440 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Black
Clear
White
Well Shape:
Rounded Square
Triangular, Flat Bottom (F-well)
Unit Size:
Pack of 100
Pack of 90
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Well Shape unitSize
13555440 Black Rounded Square Pack of 100
11947954 Clear Rounded Square Pack of 90
13545440 White Rounded Square Pack of 100
13565440 Clear Triangular, Flat Bottom (F-well) Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 13555440
Only null left
Add to basket
Add to basket

Process four times as many samples as standard 96-well plates

Optimized for robotic instruments, plate readers and liquid handling systems.

  • Highly specified polystyrene
  • Ideal for low-volume assays such as colorimetric, enzymatic, cloning, replication, and array, as well as storage
  • Flat well bottom surface provides greater area for binding
  • Working volume: 10–100μL
  • 384-well microplates conserve samples and reagents, providing cost savings and greater screening productivity
  • Untreated
  • Non-sterile

Informations sur la commande

Accommodate 12-565-511 lid (sold separately).

TRUSTED_SUSTAINABILITY

Spécification

Format Standard
Lid Without Lid
Array 16 x 24
Color Black
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 10 to 100 μL
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Rounded Square
For Use With (Application) For low volume assays such as colorimetric, enzymatic, cloning, replication and array, as well as storage
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.