missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Spacers for FB-GC10-1 Vertical Gel Caster

Spacers for use with FB-GC10-1 Vertical Gel Caster.

Brand:  Fisherbrand™

Code : 04

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11903075

  • £20.35 / Pack of 2
Estimated Shipment: 25-07-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Owl P7-CST Multiple Gradient Gel Caster
10 x 10 cm
0.05 in.
3.9 in.
5.9 in.
0.05 cm
10 cm
0.15 cm
Spacer Set
Two side spacers and one bottom spacer
1.5 cm
10 cm
3.9 in.
15 cm
0.06 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers