missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ DNA Sequencing Systems: Glass Plates for Model FB-SEQ-3545
Shop All Thermo Scientific Products
Click to view available options
Product Type:
Blank Glass
Notched Glass
Unit Size:
Each
Description
For use with model FB-SEQ-3545
Specifications
Specifications
| For Use With (Equipment) | Sequencing gel casting systems |
| Length (Metric) | 45 cm |
| Product Type | Blank Glass |
| Thickness (Metric) Teeth | 4.5 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction