Learn More
Fisherbrand™ 14-Tooth Microplate Format Combs for Horizontal Midigel Systems
14-Tooth Microplate Format Combs for Horizontal Midigel Systems
Brand:  Fisherbrand™ A1-MTD
 View more versions of this product
Product Code. 12453647
  • £50.35 / Each
Estimated Shipment: 29-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Accessory for use with Horizontal Midigel Systems
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Owl A1 Large Gel Electrophoresis System
Microplate
0.15 cm
0.28 in.
Standard Comb
14
50 μL
0.72 cm