missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Permount™ Adhesive

Product Code. 12377369 Shop All Fisher Chemical Products
Click to view available options
Volume (Metric):
100 mL
Unit Size:
Pack of 6
This item is not returnable. View return policy

Product Code. 12377369

Brand: Fisherbrand™ SP15100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cements cover glass to microscope slide.

Permount is clear with permanent adhering qualities. Consists of a synthetic polymer dissolved in toluene. Superior to both Canada balsam and gum damar for mounting specimens. 100mL container.

  • Thinner than commonly used balsam and damar solutions and easier to apply
  • Has little tendency to trap bubbles beneath cover glass
  • 115°C Melting point makes it more suitable for microprojection work
  • Refractive index is slightly higher than that of Canada balsam
  • Can be used with either artificial light or daylight
  • Absolutely neutral; will not become acidic or discolor with age

TRUSTED_SUSTAINABILITY

Specifications

Applications Can be used with either Artificial light or Daylight
Volume (English) 3.38 oz.
Volume (Metric) 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.