Learn More
OHAUS™ Frontier™ 5306 Mini Centrifuge Accessory, Rotors
Product Code. 15896612 Shop All Ohaus Products
Change view
Click to view available options
Capacity:
4 x 0.2 mL PCR8
8 x 1.5/2 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
15896612 8 x 1.5/2 mL Each
15806622 4 x 0.2 mL PCR8 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15896612 Supplier OHAUS™ Supplier No. 30129559
  • ONLINE EXCLUSIVE PRICE
    £33.00
    £32.61 / Pack of 1
    Save £0.39
    1% Off
 Restricted Item
In Stock
Add to basket
Add to basket
For use with Frontier 5306 Mini Centrifuge
Frontier rotors extend the functionality of Frontier centrifuges by increasing the number of tubes of varied types and sizes that can be used in conjunction with each of the centrifuge models. Easy to clean and operate, the rotors are durably constructed to withstand demanding centrifugation.
TRUSTED_SUSTAINABILITY
Specifications
Description Rotor, 1.5/2ml x8, FC5306
Material Polypropylene
Type Rotor
Capacity 8 x 1.5/2 mL
Max. RCF 38,800 x g
Max. Speed 6,000 rpm
For Use With (Equipment) Frontier 5306 Mini Centrifuge
Angle 45°
Includes Angle rotor 8 x 1.5/2.0mL
Biocontainment Yes, when used with biocontainment lids