Learn More
Fisherbrand™ Electrode Connection Adapters - Mercury - Free
Description
Adapters connect electrodes to various input jacks and are mercury free
- Adapters connect electrodes to various input jacks
Specifications
Specifications
| Connector Types | US Standard/Pin Plug to Radiometer Type 7 |
| For Use With (Equipment) | Electrode |
| Type | Connection Adapter |
| For Use With (Application) | Adapts electrode with U.S. Standard/Pin plugs to Radiometer Type 7 input jack |