missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ YPD Broth (Powder/Molecular Genetics)

Product Code. 10205163
Change view
Click to view available options
Quantity:
10,000 g
100 g
2000 g
500 g
Unit Size:
100g
10kg
2kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10205163 100 g 100g
10306513 500 g 500g
10797535 10,000 g 10kg
10215163 2000 g 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10205163 Supplier Fisher BioReagents™ Supplier No. BP2469100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Content And Storage 4°C
Form Powder
Grade Molecular genetics
Media Type YPD Broth
Product Type Microbiology Media
Packaging Type Poly Pail
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.