missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Tryptone (Granulated)

Product Code. 11365982
Change view
Click to view available options
Quantity:
500 g
2 kg
Packaging Type:
Poly Bottle
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Type unitSize
11365982 500 g Poly Bottle 500g
11385982 2 kg Poly Bottle 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11365982 Supplier Fisher BioReagents™ Supplier No. BP9726500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

An enzymatic digest of casein, a milk protein

  • pH (at 25°C, 2% Solution): 6.5 to 7.3

TRUSTED_SUSTAINABILITY

Specifications

Form Granulated Powder
Media Type Tryptone
Product Type Microbiology Media
Formulation Total Nitrogen ≥10%, Amino Nitrogen ≥3.5%
Packaging Type Poly Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.