missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Terrific Broth (Powder/Molecular Genetics)
Enriched medium used with glycerol in cultivating recombinant strains of Escherichia coli.
Brand: Fisher BioReagents™ BP2468-10
Code : 32
Additional Details : Weight : 10.05000kg
Description
Formulation Usage: 50.8g/L
Specifications
4°C | |
Molecular genetics | |
Microbiology Media | |
Poly Pail |
Powder | |
Terrific Broth | |
Cultivation of recombinant strains of Escherichia coli | |
10,000 g |