missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ SurePrep™ Water RNA/DNA Purification Kit

Product Code. 10711314
Change view
Click to view available options
Quantity:
25 Preps
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10711314 25 Preps Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10711314 Supplier Fisher BioReagents™ Supplier No. BP281425
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Lysis solution (15mL); Enzyme incubation buffer (4mL); Nucleic acid elution buffer (4mL); Nucleic acid wash solution (11mL); Spin columns (25); Collection tubes (25); Elution tubes, 1.7mL (25); Bead tubes (25); Filter column, 0.45μm (25)

Provides a rapid method for the isolation and purification of total RNA and DNA simultaneously from all microorganisms found in environmental water samples, including bacteria, fungi, and algae

  • Suitable for use with small and large samples of water from a variety of sources, including ocean, pond, drinking water, and wastewater
  • Optimized column, solution components, and user friendly protocol are provided for simultaneous isolation and purification of DNA and RNA
  • All RNA species are isolated, in size ranges from large mRNA and ribosomal RNA to microRNA and siRNA
  • Filter technology is used for the collection of microorganisms from water samples
  • “Bead-beating” procedure helps break open the cells and allows for efficient lysis without the use of chemicals
  • The same spin column can be used to simultaneously isolate both DNA and RNA or one nucleic acid, based on preference
  • No additional step to concentrate RNA and DNA prior to isolation
  • Rapid purification process is completed in 45 minutes
  • No special equipment or hazardous chemicals are required
  • High quality and high integrity DNA and RNA can be used for numerous downstream applications, including PCR, RT-PCR, QRT-PCR, Southern blotting, and Northern blotting

TRUSTED_SUSTAINABILITY

Specifications

Format Spin Column, Filter Column
For Use With (Application) PCR, RT-PCR, QRT-PCR, southern blotting and northern blotting
Includes Lysis solution 15mL, Enzyme incubation Buffer 4mL, Nucleic Acid Elution Buffer 4mL, Nucleic Acid wash solution 11mL, spin columns 25, collection tubes 25, 25 elution tubes, 1.7mL, bead tubes 25, 25 filter column, 0.45μm
Quantity 25 Preps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.