missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ SurePrep™ TrueTotal™ RNA Purification Kit

Product Code. 10573155
Click to view available options
Quantity:
50 Preps
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 10573155

Brand: Fisher BioReagents™ BP280050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Lysis solution (40mL), Wash solution (22mL), Elution buffer (6mL), Micro spin columns (50), Collection tubes (50), Elution tubes (1.7mL) (50)

Rapidly isolates and purifies total RNA from a variety of biological sources

  • Fast and easy processing using rapid spin-column format
  • Isolates all sizes of RNA including large RNA and small RNA species
  • No phenol or chloroform extractions
  • Isolates and detects RNA from as little as a single animal cell
  • Purifies total RNA from a variety of biological sources such as cultured animal cells, tissue samples, blood, bacteria, yeast, fungi and plants

Applications:

Purified intact RNA is suitable for Quantitative RT-PCR, Northern blotting, RNase protection, and expression array assays

TRUSTED_SUSTAINABILITY

Specifications

Format Spin Column
For Use With (Application) Quantitative RT-PCR, northern blotting, RNase protection and expression array assays
Includes Lysis solution 40mL, wash solution 22mL, Elution Buffer 6mL, micro spin columns 50, collection tubes 50, 50 elution tubes 1.7mL
Quantity 50 Preps
Yield ≈0.5μg RNA/1 x 107 cells
compliance-icons compliance-icons
Product Identifier
  • Sureprep TrueTotal RNA Purification Kit
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 C
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contains : Thiocyanic acid compound with guanidine(1:1)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.