missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ SurePrep™ Small RNA Purification Kit

Product Code. 10295253
Click to view available options
Quantity:
25 Preps
Unit Size:
Pack of 25
This item is not returnable. View return policy

Product Code. 10295253

Brand: Fisher BioReagents™ BP280125

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Lysis solution (2 x 20mL), Wash solution (22mL), RNA elution buffer (6mL), Large RNA removal columns (25), Small RNA enrichment columns (25), Collection tubes (50), Elution tubes (1.7mL) (50) and Product insert

Optimized for enrichment of small RNA by removing large RNA for studying regulation of gene expression

  • Isolate all small RNA molecules (<200 nucleotides)
  • Minimal contamination from large RNA molecules and genomic DNA
  • Fast and easy processing using rapid spin-column format
  • No phenol or chloroform extractions
  • High quality small RNA such as miRNA and siRNA can be used in various downstream applications

Applications:

Purified small RNA is suitable for use in RT-PCR, Northern blotting and microarray analysis

TRUSTED_SUSTAINABILITY

Specifications

Format Spin Column
For Use With (Application) RT-PCR, northern blotting and microarray analysis
Includes Lysis solution 2 x 20mL, wash solution 22mL, RNA Elution Buffer 6mL, large RNA removal columns 25, small RNA enrichment columns 25, collection tubes 50, elution tubes 1.7mL 50 and product insert
Quantity 25 Preps
Yield 15μg HeLa cells (1 x 106 cells), 50μg E. coli (1 x 109 cells)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.