missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ SurePrep™ RNA Cleanup and Concentration Kit

Product Code. 10609173
Click to view available options
Quantity:
50 Preps
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 10609173

Brand: Fisher BioReagents™ BP280950

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Binding solution (2 x 20mL), Wash solution (22mL), RNA elution buffer (6mL), Collection tubes (50), Spin columns (50), Elution tubes (1.7mL) (50), Product insert

Provides a rapid method to purify and concentrate RNA isolated using other methods such as phenol:chloroform extraction or after enzymatic reactions

  • Fast and easy processing using rapid spin-column format
  • Suitable for all sizes of RNA, from large mRNA to microRNA (miRNA)
  • Able to clean up RNA isolated using different methods, including phenol/chloroform extractions
  • Efficient RNA cleanup from various enzymatic reactions such as labeling, DNase treatment and in vitro transcription

Purified RNA is suitable for Quantitative RT-PCR, Northern blotting, RNase protection, primer extension and expression array assays

TRUSTED_SUSTAINABILITY

Specifications

Format Spin Column
For Use With (Application) RT-PCR, northern blotting, RNase protection, primer extension and expression array assays
Includes Binding solution 2 x 20mL, wash solution 22mL, RNA Elution Buffer 6mL, collection tubes, spin columns, elution tubes 1.7mL, product insert
Quantity 50 Preps
Yield 15μg RNA HeLa cells (1 x 106 cells), 8μg DNA HeLa cells (1 x 106 cells), 150μg Protein HeLa cells (1 x 106 cells)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.