missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Ribonuclease A (From Bovine Pancreas)
Click to view available options
Quantity:
250 mg (≥50 U/mg)
Unit Size:
250mg
Specifications
Specifications
| Color | Off-White |
| Source | Bovine pancreas |
| Quantity | 250 mg (≥50 U/mg) |
| Product Type | Ribonuclease A |
| Form | Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction