missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Restriction Enzymes - Sca I (Source: Streptomyces caespitosus)

Product Code. 11468543
Click to view available options
Quantity:
1000 U
Unit Size:
1000 units
This item is not returnable. View return policy

Product Code. 11468543

Brand: Fisher BioReagents™ BP34221

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

  • Application: Sca I restriction endonuclease digests dsDNA with sequence indicated
  • Storage Buffer Components: 10mM Tris-HCl (pH 7.4), 300mM KCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA and 50% glycerol
  • Storage: -20°C

† Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 8 to 12 U/μL
Quantity 1000 U
Cut Site AGT.ACT
Product Type ScaI
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.