missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Restriction Enzymes - Nsi I (Source: Neisseria sicca)

Product Code. 11478433
Change view
Click to view available options
Quantity:
250 U
Unit Size:
250 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478433 250 U 250 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11478433 Supplier Fisher BioReagents™ Supplier No. BP34021
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

  • Application: Nsi I restriction endonuclease digests dsDNA with sequence indicated below
  • Storage Buffer Components: 10mM Tris-HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA and 50% glycerol
  • Qualified for Blue/White Cloning Assays
  • Storage: -20°C

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Quantity 250 U
Cut Site ATGCA.T
Product Type NsiI
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.