missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Restriction Enzymes - Bcl I (Source: Bacillus caldolyticus)
Click to view available options
Quantity:
1000 units
Unit Size:
1000 units
Description
•Application: Bcl I restriction endonuclease digests dsDNA with sequence indicated below
•Storage Buffer Components: 10mM Tris-HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA and 50% glycerol
•Storage: -20°C
‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.
Specifications
Specifications
| Concentration | 10 U/μL |
| Quantity | 1000 units |
| Cut Site | T.GATCA |
| Product Type | BcII |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction