missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Peptone (Granulated)

Product Code. 11395982
Change view
Click to view available options
Quantity:
500 g
2 kg
Packaging Type:
Poly Bottle
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Type unitSize
11395982 2 kg Poly Bottle 2kg
11375982 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11395982 Supplier Fisher BioReagents™ Supplier No. BP97252
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

  • Mixture of peptides and free amino acids from pancreatic hydrolysis of animal tissue
  • pH (at 25°C, 2% Solution): 6.5 to 7.5
  • CAS: 73049-73-7

TRUSTED_SUSTAINABILITY

Specifications

Form Granulated Powder
Grade Molecular genetics
Media Type Peptone
Product Type Peptone
Formulation Total Nitrogen <10%, Amino Nitrogen <3.5%
Packaging Type Poly Bottle
Quantity 2 kg
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.