missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ OptiGrow™ Preweighed Media

Product Code. 11492964
Change view
Click to view available options
Form:
Liquid
Solid
Product Type:
Agar
Microbiology Media
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Form Product Type unitSize
11492964 Solid Microbiology Media Each
11894674 Liquid Microbiology Media Each
11820715 Solid Microbiology Media Each
11835445 Solid Agar Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11492964 Supplier Fisher BioReagents™ Supplier No. BP246810P1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Optimized for superior yield of bacterial culture

Four classic media, in convenient, preweighed pouches.

  • Preblended growth-promoting components save time and preparation costs
  • Reduce formulation errors
  • Each pouch contains enough material for 1 liter of liquid media
  • No more weighing of dusty powders
  • High quality—assured by stringent testing procedures

  • LB is used for the culturing of E. coli in molecular biology procedures
  • Terrific Broth is a buffered enriched medium used to obtain high yields of plasmid DNA from transformed E. coli

Order Info

Supplied as 10 pouches per box.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Form Solid
Grade Molecular genetics
Media Type Terrific Broth
Product Type Microbiology Media
For Use With (Application) Used for obtain high yields of plasmid DNA from transformed Escherichia coli
Packaging Type Pouch
Quantity 10 Pouches
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.