missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ NZY Broth (Powder/Molecular Genetics)

Product Code. 11428443
Change view
Click to view available options
Quantity:
100 g
2 kg
10 kg
Packaging Type:
Poly Bottle
Poly Pail
Unit Size:
100g
10kg
2kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Type unitSize
11428443 2 kg Poly Bottle 2kg
10122913 100 g Poly Bottle 100g
10002864 10 kg Poly Pail 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428443 Supplier Fisher BioReagents™ Supplier No. BP24652
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Content And Storage Keep refrigerated. (Store below 4°C/39°F.) Store in a cool, dry, well-ventilated area.
Form Powder
Grade Molecular genetics
Media Type NZY Broth
Product Type Microbiology Media
Packaging Type Poly Bottle
Quantity 2 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.