missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Nucleotides
Description
- Functionally tested in PCR
- Can be used in dideoxy sequencing, mutagenesis, and cDNA synthesis applications
- DNase-, RNase-, and protease-free
- PCR dNTP Mix is a solution of dATP, dCTP, dGTP, and dTTP
- dNTP Set contains one tube each of dATP, dCTP, dGTP, and dTTP (4 tubes/set)
Specifications
Specifications
| Format | Liquid |
| pH | 7 to 7.5 |
| Product Type | dNTP Set |
| Concentration | 100 mM |
| Purity | >98% |
| Quantity | 400 μL |
| Color | Colorless |
| For Use With (Application) | Dideoxy sequencing, mutagenesis and cDNA synthesis |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?