missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Nucleotides

Product Code. 10152863
Change view
Click to view available options
Product Type:
dATP
dCTP
dGTP
dNTP Set
Quantity:
100 μL
400 μL
Unit Size:
1 set
400µL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Quantity unitSize
10152863 dGTP 400 μL 400µL
10658973 dATP 400 μL 400µL
10410274 dCTP 400 μL 400µL
10336653 dNTP Set 100 μL 1 set
10234683 dNTP Set 400 μL 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10152863 Supplier Fisher BioReagents™ Supplier No. BP25614
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Nucleotides for PCR with a purity level of >98% triphosphate

  • Functionally tested in PCR
  • Can be used in dideoxy sequencing, mutagenesis, and cDNA synthesis applications
  • DNase-, RNase-, and protease-free
  • PCR dNTP Mix is a solution of dATP, dCTP, dGTP, and dTTP
  • dNTP Set contains one tube each of dATP, dCTP, dGTP, and dTTP (4 tubes/set)

TRUSTED_SUSTAINABILITY

Specifications

Format Liquid
pH 7 to 7.5
Molecular Weight (g/mol) 595.1
Product Type dGTP
Concentration 100 mM
Purity >98%
Quantity 400 μL
Color Colorless
For Use With (Application) Dideoxy sequencing, mutagenesis and cDNA synthesis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.