Learn More
Fisher BioReagents™ Nucleotides
Nucleotides for PCR with a purity level of >98% triphosphate
Brand:  Fisher BioReagents™ BP2560-4
Code : 32
Additional Details : CAS Number : 1927-31-7 Weight : 0.00040kg
 View more versions of this product
Product Code. 10658973
  • £147.00 / 400µL
Estimated Shipment: 16-07-2024
to see stock.
Description
Description
  • Functionally tested in PCR
  • Can be used in dideoxy sequencing, mutagenesis, and cDNA synthesis applications
  • DNase-, RNase-, and protease-free
  • PCR dNTP Mix is a solution of dATP, dCTP, dGTP, and dTTP
  • dNTP Set contains one tube each of dATP, dCTP, dGTP, and dTTP (4 tubes/set)
Specifications
Specifications
Liquid
579.2
100 mM
400 μL
Dideoxy sequencing, mutagenesis and cDNA synthesis
7 to 7.5
dATP
>98%
Colorless
Product Suggestions
Product Suggestions
Videos
SDS Documents