missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Microbiology Media Additives: Tryptone
Additive for preparing bacterial nutrient media
£125.72 - £672.04
Specifications
Form | Dehydrated Powder |
---|---|
Grade | Molecular genetics |
Media Type | Tryptone |
Product Type | Microbiology Media |
For Use With (Application) | Pancreatic digest of casein is used as a nitrogen source for bacteria |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12787099
|
Fisher BioReagents™
12787099 |
2000 g |
£672.04
2kg |
In Stock
Log in to see stock. |
|||||
12797099
|
Fisher BioReagents™
12797099 |
500 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Note: This is a product of animal origin (pancreas).Specifications
Dehydrated Powder | |
Tryptone | |
Pancreatic digest of casein is used as a nitrogen source for bacteria |
Molecular genetics | |
Microbiology Media | |
Poly Bottle |