missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Microbiology Media Additives: Tryptone

Product Code. 12797099
Change view
Click to view available options
Quantity:
500 g
2000 g
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12797099 500 g 500g
12787099 2000 g 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12797099 Supplier Fisher BioReagents™ Supplier No. 12797099
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Additive for preparing bacterial nutrient media

Note: This is a product of animal origin (pancreas).

Specifications

Description Bacterial Culture
Form Dehydrated Powder
Grade Molecular genetics
Media Type Tryptone
Product Type Dehydrated Culture Media
For Use With (Application) Pancreatic digest of casein is used as a nitrogen source for bacteria
Packaging Type Poly Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.