missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Microbiology Media Additives: Tryptone
Description
Note: This is a product of animal origin (pancreas).
Specifications
Specifications
| Form | Dehydrated Powder |
| Grade | Molecular genetics |
| Media Type | Tryptone |
| Product Type | Microbiology Media |
| For Use With (Application) | Pancreatic digest of casein is used as a nitrogen source for bacteria |
| Packaging Type | Poly Bottle |
| Quantity | 2000 g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?