missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Microbiology Media Additives: Peptone
Description
Dehydrated.
Specifications
Specifications
| Form | Powder |
| Grade | Molecular genetics |
| Media Type | Peptone |
| Product Type | Media Additive |
| For Use With (Application) | Nitrogen in a form readily available to bacteria |
| Packaging Type | Poly Bottle |
| Quantity | 2000 g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?