missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Fisher BioReagents™ Microbiology Media Additives: Acid Casein Peptone

Product Code. 11493853
Change view
Click to view available options
Quantity:
2000 g
Packaging Type:
Poly Pail
Unit Size:
2kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Type unitSize
11493853 2000 g Poly Pail 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11493853 Supplier Fisher BioReagents™ Supplier No. 11493853

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Additive for preparing bacterial nutrient media

Acid Casein Peptone

Dehydrated.

Specifications

Form Powder
Grade Molecular genetics
Product Type Casamino Acid
For Use With (Application) Acid hydrolyzed casein's low concentrations of sodium chloride and iron permit toxin production
Packaging Type Poly Pail
Quantity 2000 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.