missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Lysozyme, Egg White (White Cryst. Powder)

Product Code. 10421545
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Unit Size:
10g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10421545 10 g 10g
10536394 1 g 1g
10535054 5 g 5g
10421735 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10421545 Supplier Fisher BioReagents™ Supplier No. BP53510
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
  • Application: This enzyme is used to lyse E. coli for preparation of plasmid DNA from transformed cell cultures
  • Not on TSCA inventory: for research and development use only; not for manufacturing or commercial purposes
  • Enzyme Activity: ≈20,000U/mg dry weight
  • Solubility: Readily soluble at 5mg/mL in 0.1M KH2PO4 to give colorless solution
TRUSTED_SUSTAINABILITY

Specifications

Color White
For Use With (Application) Used to lyse E. coli for preparation of plasmid DNA from transformed cell cultures
Content And Storage <0°C
Source Egg white
Quantity 10 g
Product Type Lysozyme
Form Crystalline Powder
compliance-icons
Product Identifier
  • Lysozyme, egg white
Signal Word
  • Danger
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.