Learn More
Fisher BioReagents™ Lysozyme, Egg White (White Cryst. Powder)
Brand: Fisher BioReagents™ BP535-25
Code : A0
Additional Details : CAS Number : 12650-88-3 Weight : 0.08000kg
Description
- Application: This enzyme is used to lyse E. coli for preparation of plasmid DNA from transformed cell cultures
- Not on TSCA inventory: for research and development use only; not for manufacturing or commercial purposes
- Enzyme Activity: ≈20,000U/mg dry weight
- Solubility: Readily soluble at 5mg/mL in 0.1M KH2PO4 to give colorless solution
Specifications
White | |
<0°C | |
25 g | |
Crystalline Powder |
Used to lyse E. coli for preparation of plasmid DNA from transformed cell cultures | |
Egg white | |
Lysozyme |
Safety and Handling
- Lysozyme, egg white
Signal Word
- Danger
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .