missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Leupeptin Hemisulfate
Description
(Acetyl-Leu-Leu-Arg-al), Valeramide
A biochemical used as an inhibitor for trypsin, plasmin, papain, and cathepsin B.
Specifications
Specifications
| Color | White |
| For Use With (Application) | Inhibitor for Trypsin, Plasmin, Papain, Cathepsin B |
| Content And Storage | -20°C |
| Quantity | 25 mg |
| Product Type | Leupeptin Hemisulfate |
| Molecular Weight (g/mol) | 475.60 |
| Form | Chunks, Powder |
Safety and Handling
Product Identifier
- Leupeptin Hemisulfate
- Warning
- Acute toxicity Category 4
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?