missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Leupeptin Hemisulfate
Click to view available options
Quantity:
1 mg
5 mg
25 mg
100 mg
Unit Size:
100mg
1mg
25mg
5mg
Description
(Acetyl-Leu-Leu-Arg-al), Valeramide
A biochemical used as an inhibitor for trypsin, plasmin, papain, and cathepsin B.
Specifications
Specifications
| Color | White |
| For Use With (Application) | Inhibitor for Trypsin, Plasmin, Papain, Cathepsin B |
| Content And Storage | -20°C |
| Quantity | 100 mg |
| Product Type | Leupeptin Hemisulfate |
| Molecular Weight (g/mol) | 475.60 |
| Form | Chunks, Powder |
Safety and Handling
Product Identifier
- Leupeptin Hemisulfate
- Warning
- Acute toxicity Category 4
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction