missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ LB Agar, Miller (Pre-Buffered Capsules)

Product Code. 11366002
Change view
Click to view available options
Quantity:
25 x 20 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11366002 25 x 20 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11366002 Supplier Fisher BioReagents™ Supplier No. BP9734500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.

  • Directions for Media Preparation: Add 20 capsules per 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water). Heat to completely dissolve capsule. Before use, sterilize by autoclaving for 15 minutes.
  • Product is in capsule form
  • Each capsule contains 1g of broth
  • Capsules are prebuffered and set to pH 7.2 at 20°C using a 10mM solution of tris/tris HCl
  • pH will vary with temperature
TRUSTED_SUSTAINABILITY

Specifications

Format Pre-Buffered Capsules
Media Type LB Agar
Product Type Agar
Formulation Casein Peptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L, Agar 15g/L, TRIS/TRIS HCl 1.5g/L
For Use With (Application) For maintenance and propagation of Escherichia coli in molecular biology procedures
Packaging Type Poly Bottle
Quantity 25 x 20 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.