missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Taq DNA Polymerase

Product Code. 10206744
Change view
Click to view available options
Composition:
10X assay buffer A : Tris-HCl 100mM, pH 9 at 25°C, KCl 500mM and MgCl2 15mM solution in a separate vial
10X assay buffer B : Tris-HCl 100mM, pH 9 at 25°C, KCl 500mM and MgCl2 25mM solution in a separate vial
Quantity:
100 Units
25 × 100 Units
2500 Units
5 × 100 Units
5 × 2500 Units
5 × 500 Units
500 Units
Unit Size:
100 units
2500 units
500 units
Pack of 5
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Composition Quantity unitSize
10206744 10X assay buffer B : Tris-HCl 100mM, pH 9 at 25°C, KCl 500mM and MgCl2 25mM solution in a separate vial 25 × 100 Units 2500 units
10458923 10X assay buffer A : Tris-HCl 100mM, pH 9 at 25°C, KCl 500mM and MgCl2 15mM solution in a separate vial 5 × 500 Units 2500 units
10448733 10X assay buffer A : Tris-HCl 100mM, pH 9 at 25°C, KCl 500mM and MgCl2 15mM solution in a separate vial 100 Units 100 units
10717185 10X assay buffer A : Tris-HCl 100mM, pH 9 at 25°C, KCl 500mM and MgCl2 15mM solution in a separate vial 25 × 100 Units 2500 units
10091263 10X assay buffer A : Tris-HCl 100mM, pH 9 at 25°C, KCl 500mM and MgCl2 15mM solution in a separate vial 5 × 100 Units 500 units
10458733 10X assay buffer A : Tris-HCl 100mM, pH 9 at 25°C, KCl 500mM and MgCl2 15mM solution in a separate vial 500 Units 500 units
10698393 10X assay buffer A : Tris-HCl 100mM, pH 9 at 25°C, KCl 500mM and MgCl2 15mM solution in a separate vial 2500 Units 2500 units
10144924 10X assay buffer A : Tris-HCl 100mM, pH 9 at 25°C, KCl 500mM and MgCl2 15mM solution in a separate vial 5 × 2500 Units Pack of 5
10449693 10X assay buffer B : Tris-HCl 100mM, pH 9 at 25°C, KCl 500mM and MgCl2 25mM solution in a separate vial 100 Units 100 units
10346363 10X assay buffer B : Tris-HCl 100mM, pH 9 at 25°C, KCl 500mM and MgCl2 25mM solution in a separate vial 5 × 100 Units 500 units
10216744 10X assay buffer B : Tris-HCl 100mM, pH 9 at 25°C, KCl 500mM and MgCl2 25mM solution in a separate vial 2500 Units 2500 units
10356363 10X assay buffer B : Tris-HCl 100mM, pH 9 at 25°C, KCl 500mM and MgCl2 25mM solution in a separate vial 500 Units 500 units
10104974 10X assay buffer B : Tris-HCl 100mM, pH 9 at 25°C, KCl 500mM and MgCl2 25mM solution in a separate vial 5 × 2500 Units Pack of 5
10071553 10X assay buffer B : Tris-HCl 100mM, pH 9 at 25°C, KCl 500mM and MgCl2 25mM solution in a separate vial 5 × 500 Units 2500 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Product Code. 10206744 Supplier Fisher BioReagents™ Supplier No. FB600055
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Allows up to 100 cycles of amplification without loss of activity

  • Licensed for use in PCR
  • Uses a thermostable DNA polymerase that has been encoded by a modified form of Thermus aquaticus DNA polymerase gene that has been inserted into an E. coli host
  • Has a 5«→3« exonuclease activity
  • Available with two different reaction buffers
  • Buffer A contains an optimized final concentration of 15mM MgCl2 for most PCR amplifications
  • Buffer B does not contain MgCl2; MgCl2 is provided in a separate vial as a 25mM solution
  • Storage Buffer: 20mM Tris-HCl, pH 7.5 (at room temperature), 100mM KCl, 0.1mM EDTA, 1mM DTT, 50% Glycerol, 0.5% Tween™ 20, 0.5% Nonidet P40™
  • Recommended Assay Buffer: 10X Assay Buffer A (100mM Tris-HCl, pH 9.0 [at 25°C/77°F]; 500mM KCl; 15mM MgCl2) or 10X Assay Buffer B (100mM Tris-HCl, pH 9.0 [at 25°C/77°F]; 500mM KCl) and 25mM MgCl2 solution in a separate vial

Unit Definition: One unit of enzyme is defined as the amount that will incorporate 10nmol of dNTPs into acid-insoluble material per 30 minutes in a 10-minute incubation at 74°C (165°F) under the assay conditions above

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Composition 10X assay buffer B : Tris-HCl 100mM, pH 9 at 25°C, KCl 500mM and MgCl2 25mM solution in a separate vial
Content And Storage -20°C
Description Taq DNA Polymerase
Form Liquid
Format Poly Tube
Quantity 25 × 100 Units
For Use With (Application) Standard PCR
Storage Buffer Tris-HCl 20mM, pH 7.5 (at room temperature), KCl 100mM, EDTA 0.1mM, DTT 1mM, Glycerol 50%, Tween™ 20 0.5%, Nonidet™ P40™ 0.5%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.