missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ exACTGene™ dNTPs
Description
- >99% triphosphate
- Chemically synthesized to meet a high purity standard
- Ensure consistent results and optimum sensitivity for many rigorous applications
- Tested for purity by HPLC, performance in long-template PCR, and absence of DNase, RNase, and protease activity to ensure dependable performance and lot-to-lot consistency
- Ultrapure dNTPs will enhance results for the following procedures: QPCR, high-fidelity PCR, long template (>8kb) PCR, RT-PCR, fluorescent sequencing, cDNA synthesis, Pyrosequencing™, and in vitro mutagenesis
Specifications
Specifications
| Format | Liquid |
| pH | 7.5 |
| Product Type | dCTP |
| Content And Storage | -20°C |
| Concentration | 100 mM |
| Purity | >99% |
| Quantity | 250 μL |
| Color | Colorless |
| For Use With (Application) | PCR, DNA labeling and DNA sequencing |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?