missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ exACTGene™ DNA Ladders

Product Code. 10122823
Click to view available options
No. of Bands:
8
10
11
12
13
14
15
16
19
For Use With (Application):
Clone Identification
Fast run times, small size DNA
General purpose, extra large size
General purpose, large digested DNA
General purpose, small size DNA
General purpose, wide size range
Large size PCR or cloning
PCR size confirmation
Quick check of PCR or Enzyme digestion
Small and large cloning
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10122823

Brand: Fisher BioReagents™ BP2577100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ready-to-use DNA size standards are stable at room temperature—no need to thaw and refreeze

  • Specially engineered to provide blunt-end DNA of precise length for better band quality than classic markers
  • Ideal for size assessment and quantitative determination of DNA mass
  • Contain loading dye to reduce pipetting steps and save time
  • Have two or three high intensity reference bands for better sizing
  • Provided in sufficient quantity to load 100 lanes

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Product Type DNA Molecular Weight Marker/Ladder
Form Liquid
Formulation Premixed with loading dye
For Use With (Application) Small and large cloning
No. of Bands 16
Quantity 100 Lanes
Storage Requirements Stable at room temperature for two years.
Sufficient For 100 Lanes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.