missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Collagenase (From Clostridium histolyticum, Islet Isolation Grade)

Product Code. 11463094
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 11463094

Brand: Fisher BioReagents™ BP26491

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

  • Application: Collagenase is most suitable for pancreatic islet isolation
  • Unit Definition: One unit will liberate peptides from collagen equivalent in ninhydrin color to 3.0μmole of L-leucine in 18 hr at pH 7.4 and 37°C
  • Storage: 0° to 5°C, dessicate

TRUSTED_SUSTAINABILITY

Specifications

Color Yellowish Brown
pH 7.4
For Use With (Application) Suitable for pancreatic islet isolation
Content And Storage 0°C to 5°C
Source Clostridium histolyticum
Quantity 1 g
Product Type Collagenase
Molecular Weight (g/mol) 105000
Form Powder
compliance-icons
Product Identifier
  • Collagenase, Islet Isolation Grade, minium of 3500 units/g, from Clostridium histolyticum yellowish brown lyophilized powder
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.