missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ ChromoMax™ IPTG/X-Gal Solution

Product Code. 10225163
Change view
Click to view available options
Quantity:
1 mL
10 mL
Unit Size:
10mL
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10225163 10 mL 10mL
10133443 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10225163 Supplier Fisher BioReagents™ Supplier No. BP420010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Accelerate your blue/white colony screening

Ready-to-apply ChromoMax IPTG/X-Gal solution is a proprietary formulation that allows you to skip the cumbersome preparation process that traditionally requires tedious weighing and dissolving of fine powder in toxic solvents such as DMSO or DMF.

  • Premixed IPTG/X-Gal Solution
  • No weighing or dissolving of components
  • No DMSO or DMF required
  • Saves time and money
  • Dependable results

  • Can be safely handled outside of a fume hood
  • ChromoMax Solution does not freeze even when stored at -20°C, allowing for immediate use without thawing
  • Dries quickly on the plate, further accelerating the screening protocol
  • Provides fast color development and does not require additional 4°C incubation to enhance color
  • Clear colorless to yellow solution

ChromoMax/ITPTG/X-Gal Solution is suitable for use with all commercially available competent cells that utilize lacZ blue/white screening. Tested using Fisher BioReagents transformed TransMax Competent Cells resulting in clearly differentiated blue and white colonies.

Order Info

The 50mL volume is conveniently packaged in five 10mL bottles, preventing accidental contamination of large volume stock.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Additive IPTG/X-Gal Solution
Form Liquid
For Use With (Application) All commercially available competent cells that utilize lacZ blue/white screening
Packaging Type Bottle
Quantity 10 mL
compliance-icons compliance-icons
Product Identifier
  • ChromoMax
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H360D-May damage the unborn child.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.