missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fish Canyon Tuff Creede, CO (Dacite), CRM, ARMI∣MBH
Description
Fish Canyon Tuff Creede, CO (Dacite), CRM
Specifications
Specifications
| Chemical Name or Material | Chromium/Nickel/Molybdenum/Tungsten/Thallium/Antimony/Thulium/Bismuth/Cadmium/Silver/Indium/Manganese/Phosphorus/Manganese(II) oxide/Phosphorus pentoxide/Lutetium/Titanium/Titanium dioxide/Terbium/Holmium/Magnesium/Tantalum/Loss on ignition/Europium/Magnesium oxide/Tin/Cesium/Beryllium/Ytterbium/Erbium/Calcium/Sodium/Potassium/Arsenic/Iron/Calcium oxide/Uranium/Sodium oxide/Potassium oxide/Dysprosium/Gadolinium/Total Iron expressed as Fe2O3/Hafnium/Samarium/Scandium/Aluminium/Copper/Praseodymium/Cobalt/Thorium/Niobium/Lead/Aluminium oxide/Gallium/Lithium/Yttrium/Silicon/Neodymium/Lanthanum/Silicon dioxide/Zinc/Vanadium/Cerium/Rubidium/Zirconium/Strontium/Barium |
| CAS | 14808-60-7 |
| Quantity | 75 g |
| Form | Fine Powder |
| Concentration or Composition (by Analyte or Components) | Chromium (5) mg/kg/Nickel (4) mg/kg/Molybdenum (0.7) mg/kg/Tungsten (0.6) mg/kg/Thallium (0.4) mg/kg/Antimony (0.3) mg/kg/Thulium (0.3) mg/kg/Bismuth (0.1) mg/kg/Cadmium (0.07) mg/kg/Silver (0.05) mg/kg/Indium (0.04) mg/kg/Manganese 0.081 %/Phosphorus 0.098 %/Manganese(II) oxide 0.104 %/Phosphorus pentoxide 0.225 %/Lutetium 0.33 mg/kg/Titanium 0.344 %/Titanium dioxide 0.573 %/Terbium 0.66 mg/kg/Holmium 0.74 mg/kg/Magnesium 0.84 %/Tantalum 1.02 mg/kg/Loss on ignition 1.2 %/Europium 1.33 mg/kg/Magnesium oxide 1.4 %/Tin 1.7 mg/kg/Cesium 2 mg/kg/Beryllium 2.09 mg/kg/Ytterbium 2.2 mg/kg/Erbium 2.22 mg/kg/Calcium 2.55 %/Sodium 2.72 %/Potassium 3.2 %/Arsenic 3.3 mg/kg/Iron 3.32 %/Calcium oxide 3.57 %/Uranium 3.6 mg/kg/Sodium oxide 3.67 %/Potassium oxide 3.86 %/Dysprosium 3.9 mg/kg/Gadolinium 4.6 mg/kg/Total Iron expressed as Fe2O3 4.74 %/Hafnium 5.6 mg/kg/Samarium 6 mg/kg/Scandium 8.1 mg/kg/Aluminium 8.52 %/Copper 9 mg/kg/Praseodymium 9.5 mg/kg/Cobalt 10.3 mg/kg/Thorium 10.8 mg/kg/Niobium 15.2 mg/kg/Lead 16 mg/kg/Aluminium oxide 16.1 %/Gallium 18.5 mg/kg/Lithium 19.5 mg/kg/Yttrium 21.8 mg/kg/Silicon 30.1 %/Neodymium 34.7 mg/kg/Lanthanum 44.2 mg/kg/Silicon dioxide 64.4 %/Zinc 68 mg/kg/Vanadium 69 mg/kg/Cerium 84 mg/kg/Rubidium 104 mg/kg/Zirconium 209 mg/kg/Strontium 526 mg/kg/Barium 924 mg/kg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?