missing translation for 'onlineSavingsMsg'
Learn More

Fisetin, MedChemExpress

Product Code. 16472509
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472509 1 g 1g
16412366 500 mg 500mg
16409026 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16472509 Supplier Medchem Express Supplier No. HYN0182
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Fisetin is a natural flavonol found in many fruits and vegetables with various benefits, such as antioxidant, anticancer, neuroprotection effects.

Fisetin is a natural flavonol found in many fruits and vegetables with various benefits, such as antioxidant, anticancer, neuroprotection effects.

Specifications

Chemical Name or Material Fisetin
CAS 528-48-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H10O6
Quantity 1 g
Solubility Information DMSO : 25 mg/mL (87.34 mM; Need ultrasonic)
SMILES O=C1C(O)=C(C2=CC=C(O)C(O)=C2)OC3=CC(O)=CC=C13
Molecular Weight (g/mol) 286.24
Formula Weight 286.24
Percent Purity 96.74%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.