missing translation for 'onlineSavingsMsg'
Learn More

Firsocostat, MedChemExpress

Product Code. 16459727
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16459727

Brand: Medchem Express HY16901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Firsocostat (ND-630; GS-0976; NDI-010976) is an acetyl-CoA carboxylase (ACC) inhibitor; inhibits human ACC1 and ACC2 with IC50 values of 2.1 and 6.1 nM, respectively.

Firsocostat (ND-630

Specifications

Chemical Name or Material Firsocostat
CAS 1434635-54-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C28H31N3O8S
Quantity 5 mg
Synonym ND-630 GS-0976 NDI-010976
Solubility Information DMSO : ≥ 50 mg/mL (87.78 mM)
SMILES O=C(C(C(C)=C(C1=NC=CO1)S2)=C2N3C[C@H](OC4CCOCC4)C5=C(OC)C=CC=C5)N(C(C)(C)C(O)=O)C3=O
Molecular Weight (g/mol) 569.63
Formula Weight 569.63
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.