missing translation for 'onlineSavingsMsg'
Learn More

Firocoxib, TRC

Product Code. 30395079
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395079 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395079 Supplier Toronto Research Chemicals Supplier No. TRCF3560001MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Firocoxib
CAS 189954-96-9
Purity Grade Notes HPLC
Molecular Formula C17 H20 O5 S
Quantity 1 mg
Synonym 3-(Cyclopropylmethoxy)-5,5-dimethyl-4-[4-(methylsulfonyl)phenyl]-2(5H)-furanone, 3-(Cyclopropyl-methoxy)-5,5-dimethyl-4-(4-methylsulfonyl)phenyl)-5H-furan-2-one, Equioxx, Equixx, Firocoxib, Librens, ML 1785713, Previcox
InChI Formula InChI=1S/C17H20O5S/c1-17(2)14(12-6-8-13(9-7-12)23(3,19)20)15(16(18)22-17)21-10-11-4-5-11/h6-9,11H,4-5,10H2,1-3H3
SMILES CC1(C)OC(=O)C(=C1c2ccc(cc2)S(=O)(=O)C)OCC3CC3
IUPAC Name 3-(cyclopropylmethoxy)-5,5-dimethyl-4-(4-methylsulfonylphenyl)furan-2-one
Molecular Weight (g/mol) 336.4
Formula Weight 336.1031
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.