missing translation for 'onlineSavingsMsg'
Learn More

Firocoxib, MedChemExpress

Product Code. 16442267
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16442267

Brand: Medchem Express HY14670

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Firocoxib (ML 1785713) is a potent, selective and orally active COX-2 inhibitor with an IC50 of 0.13 μM. Firocoxib shows 58-fold more selective for COX-2 than COX-1 (IC50 of 7.5 μM). Firocoxib has anti-inflammatory effects.

Firocoxib (ML 1785713) is a potent, selective and orally active COX-2 inhibitor with an IC50 of 0.13 μM. Firocoxib shows 58-fold more selective for COX-2 than COX-1 (IC50 of 7.5 μM). Firocoxib has anti-inflammatory effects.

Specifications

Chemical Name or Material Firocoxib
CAS 189954-96-9
Color White
Purity Grade Notes Research
Molecular Formula C17H20O5S
Quantity 50 mg
Synonym ML 1785713
Solubility Information DMSO : ≥ 52 mg/mL (154.58 mM)
SMILES O=C1OC(C)(C)C(C2=CC=C(S(=O)(C)=O)C=C2)=C1OCC3CC3
Molecular Weight (g/mol) 336.4
Formula Weight 336.4
Percent Purity 98.42%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.