missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell Safety Products™ Fireman Glove
Description
- Heat resistant glove (formerly used as glove for firemen).
- Yellow water-repellent cowhide grain leather glove.
- Gunn cut, winged thumb, gauntlet type.
- Hand lined with Kevlar™ cotton jersey for comfort and protection from cuts and thermal hazards.
- Stitched with Kevlar™ thread for thermal insulation and sturdy seam resistance to heat and wear.
- Elasticated back.
- Velcro strap.
- Split leather cuff.
Specifications
Specifications
| Certifications/Compliance | CE CATEGORY III Irreversible risks, EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical hazards, EN 407-2004 Thermal hazards, EC Category PPE 2, EC Certificate Number 0075/797/162/12/09/0517 |
| Classification | Dexterity: 5 |
| Cut Style | Gunn |
| Cut Resistance Rating | 2 |
| Material | Grain Leather, Cowhide Leather, Aramid, Cotton, Dupont™ Kevlar™ Aramid Fibers |
| Product Type | Gloves |
| Size | 8 |
| Style | Gauntlet |
| Thickness (Metric) | 1.1 to 1.3 mm |
| For Use With (Application) | Army-Defense and Fire Protection industries |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?