missing translation for 'onlineSavingsMsg'
Learn More

Firategrast, MedChemExpress

Product Code. 16496268
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16496268

Brand: Medchem Express HY14951

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Firategrast (SB 683699) is an orally active and specific α4β1/α4β7 integrin antagonist. Firategrast reduces trafficking of lymphocytes into the central nervous system (CNS) and decreases multiple sclerosis (MS) activity.

Firategrast (SB 683699) is an orally active and specific α4β1/α4β7 integrin antagonist. Firategrast reduces trafficking of lymphocytes into the central nervous system (CNS) and decreases multiple sclerosis (MS) activity.

Specifications

Chemical Name or Material Firategrast
CAS 402567-16-2
Color White
Purity Grade Notes Research
Molecular Formula C27H27F2NO6
Quantity 10 mg
Synonym SB 683699
Solubility Information DMSO : ≥ 100 mg/mL (200.20 mM) ∣Ethanol : ≥ 50 mg/mL (100.10 mM)
SMILES O=C(O)[C@@H](NC(C1=C(F)C=CC=C1F)=O)CC2=CC=C(C3=C(OC)C=C(COCC)C=C3OC)C=C2
Molecular Weight (g/mol) 499.5
Formula Weight 499.5
Percent Purity 99.62%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.