missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fipronil, TRC
Specifications
Specifications
| Chemical Name or Material | Fipronil |
| CAS | 120068-37-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C12 H4 Cl2 F6 N4 O S |
| Quantity | 1 g |
| Synonym | Fipronil, 1H-Pyrazole-3-carbonitrile, 5-amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-[(trifluoromethyl)sulfinyl]-, 5-Amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-[(trifluoromethyl)sulfinyl]-1H-pyrazole-3-carbonitrile, (±)-Fipronil, 1-(2,6-Dichloro-4-trifluoromethylphenyl)-3-cyano-5-amino-4-(trifluoromethylsulfinyl)pyrazole, 1-[2,6-Dichloro-4-(trifluoromethyl)phenyl]-3-cyano-4-[(trifluoromethyl)sulfinyl]-5-aminopyrazole, 5-Amino-1-(2,6-dichloro-4-trifluoromethylphenyl)-3-cyano-4-trifluoromethylsulfinylpyrazole, Agenda, Agenda SC, Albatross (insecticide), Ascend, Blitz, Chipco Choice, Cosmos, Cosmos (insecticide), Eliminall, Fluocyanobenpyrazole, Formidor L, Frontline Spot-on, Frontline Spray, Frontline Top Spot, Goliath gel, Granedo MC, Grenade MC, Jump 80WG, Maxforce FC, Maxforce FC Select Roach Killer Bait Gel, Over'n Out, Prince, Regent, Regent 0.3G, Regent 200 SC, Regent 250 FS, Regent 800WG, Regent TS, Standak, Standak 250SC, Taj, Termidor, Termidor (insecticide), Termidor 80WG, TopChoice, Topline |
| InChI Formula | InChI=1S/C12H4Cl2F6N4OS/c13-5-1-4(11(15,16)17)2-6(14)8(5)24-10(22)9(7(3-21)23-24)26(25)12(18,19)20/h1-2H,22H2 |
| SMILES | Nc1c(c(nn1c2c(Cl)cc(cc2Cl)C(F)(F)F)C#N)S(=O)C(F)(F)F |
| IUPAC Name | 5-amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-(trifluoromethylsulfinyl)pyrazole-3-carbonitrile |
| Molecular Weight (g/mol) | 437.15 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?