missing translation for 'onlineSavingsMsg'
Learn More

Fipronil, TRC

Product Code. 30392113
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392113 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392113 Supplier Toronto Research Chemicals Supplier No. TRCF3422005G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fipronil
CAS 120068-37-3
Purity Grade Notes HPLC
Molecular Formula C12 H4 Cl2 F6 N4 O S
Quantity 5 g
Synonym Fipronil, 1H-Pyrazole-3-carbonitrile, 5-amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-[(trifluoromethyl)sulfinyl]-, 5-Amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-[(trifluoromethyl)sulfinyl]-1H-pyrazole-3-carbonitrile, (±)-Fipronil, 1-(2,6-Dichloro-4-trifluoromethylphenyl)-3-cyano-5-amino-4-(trifluoromethylsulfinyl)pyrazole, 1-[2,6-Dichloro-4-(trifluoromethyl)phenyl]-3-cyano-4-[(trifluoromethyl)sulfinyl]-5-aminopyrazole, 5-Amino-1-(2,6-dichloro-4-trifluoromethylphenyl)-3-cyano-4-trifluoromethylsulfinylpyrazole, Agenda, Agenda SC, Albatross (insecticide), Ascend, Blitz, Chipco Choice, Cosmos, Cosmos (insecticide), Eliminall, Fluocyanobenpyrazole, Formidor L, Frontline Spot-on, Frontline Spray, Frontline Top Spot, Goliath gel, Granedo MC, Grenade MC, Jump 80WG, Maxforce FC, Maxforce FC Select Roach Killer Bait Gel, Over'n Out, Prince, Regent, Regent 0.3G, Regent 200 SC, Regent 250 FS, Regent 800WG, Regent TS, Standak, Standak 250SC, Taj, Termidor, Termidor (insecticide), Termidor 80WG, TopChoice, Topline
InChI Formula InChI=1S/C12H4Cl2F6N4OS/c13-5-1-4(11(15,16)17)2-6(14)8(5)24-10(22)9(7(3-21)23-24)26(25)12(18,19)20/h1-2H,22H2
SMILES Nc1c(c(nn1c2c(Cl)cc(cc2Cl)C(F)(F)F)C#N)S(=O)C(F)(F)F
IUPAC Name 5-amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-(trifluoromethylsulfinyl)pyrazole-3-carbonitrile
Molecular Weight (g/mol) 437.15
Formula Weight 435.9387
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.